Anti-RLA, DR Mouse Monoclonal Antibody (Biotin) [clone: 5K114]

Lieferant: US Biological
R2032-50B
USBIR2032-50BEA 805 CHF
USBIR2032-50B
Anti-RLA, DR Mouse Monoclonal Antibody (Biotin) [clone: 5K114]
Antikörper
Type: Primary
Antigen: RLA, DR
Clonality: monoclonal
Clone: 5K114
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR