Anti-RIPK3 Rabbit Polyclonal Antibody

Lieferant: US Biological
R2031-74D
USBIR2031-74DEA 937 CHF
USBIR2031-74D
Anti-RIPK3 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:50-1:100; Immunochemistry: 1:10-1:50

Type: Primary
Antigen: RIPK3 (receptor-interacting serine-threonine kinase 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR