Anti-RAB17 Goat Polyclonal Antibody

Lieferant: US Biological
R0009-30
USBIR0009-30EA 899 CHF
USBIR0009-30
Anti-RAB17 Goat Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:64000

Type: Primary
Antigen: RAB17 (RAB17, member RAS oncogene family)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR