Anti-Ornithine Decarboxylase Guinea pig Polyclonal Antibody

Lieferant: US Biological
O8025-16
USBIO8025-16EA 1690 CHF
USBIO8025-16
Anti-Ornithine Decarboxylase Guinea pig Polyclonal Antibody
Antikörper
Recommended dilution: Immunofluorescence: 1:500-1:800

Type: Primary
Antigen: Ornithine Decarboxylase
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Guinea pig
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR