Anti-NMS Rabbit Polyclonal Antibody (Carboxyfluorescein)

Lieferant: US Biological
N2171-79U3
USBIN2171-79U3EA 1630 CHF
USBIN2171-79U3
Anti-NMS Rabbit Polyclonal Antibody (Carboxyfluorescein)
Antikörper
Type: Primary
Antigen: NMS (neuromedin S)
Clonality: polyclonal
Clone:
Conjugation: Carboxyfluorescein
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR