Anti-NAGLU Mouse Monoclonal Antibody [clone: 11C455]

Lieferant: US Biological
N0017-05
USBIN0017-05EA 920 CHF
USBIN0017-05
Anti-NAGLU Mouse Monoclonal Antibody [clone: 11C455]
Antikörper
Recommended dilution: Western Blot: 1:500-1:1000

Type: Primary
Antigen: NAGLU (N-acetylglucosaminidase, alpha)
Clonality: monoclonal
Clone: 11C455
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR