Anti-Myb-binding protein 1A Rabbit Polyclonal Antibody

Lieferant: US Biological
M9600-04
USBIM9600-04EA 1090 CHF
USBIM9600-04
Anti-Myb-binding protein 1A Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 1-3?g/ml

Type: Primary
Antigen: Myb-binding protein 1A
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR