Anti-Melanocortin Receptor 4 Rabbit Polyclonal Antibody (Biotin)

Lieferant: US Biological
M2868-39D1
USBIM2868-39D1EA 1180 CHF
USBIM2868-39D1
Anti-Melanocortin Receptor 4 Rabbit Polyclonal Antibody (Biotin)
Antikörper
Type: Primary
Antigen: Melanocortin Receptor 4
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR