Anti-Melanin Concentrating Hormone Rabbit Polyclonal Antibody

Lieferant: US Biological
M2867-70F
USBIM2867-70FEA 1180 CHF
USBIM2867-70F
Anti-Melanin Concentrating Hormone Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Melanin Concentrating Hormone
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR