Anti-Liver Antigen Mouse Monoclonal Antibody [clone: EP-1]

Lieferant: US Biological
L3150-03B
USBIL3150-03BEA 924 CHF
USBIL3150-03B
Anti-Liver Antigen Mouse Monoclonal Antibody [clone: EP-1]
Antikörper
Type: Primary
Antigen: Liver Antigen
Clonality: monoclonal
Clone: EP-1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR