Anti-LTF Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: US Biological
L1021-82J
USBIL1021-82JEA 1050 CHF
USBIL1021-82J
Anti-LTF Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
Recommended dilution: ELISA: 1:1000-1:7000

Type: Primary
Antigen: LTF (lactotransferrin)
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR