Anti-IL4 Rabbit Polyclonal Antibody

Lieferant: US Biological
I8426-26A-100
USBII8426-26A-100EA 597 CHF
USBII8426-26A-100
Anti-IL4 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: IL4 (interleukin 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR