Anti-INS Guinea pig Polyclonal Antibody

Lieferant: US Biological
I7656-17
USBII7656-17EA 888 CHF
USBII7656-17
Anti-INS Guinea pig Polyclonal Antibody
Antikörper
Recommended dilution: Immunofluorescence: 1:400-1:800

Type: Primary
Antigen: INS (insulin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Guinea pig
Isotype: IgG
Reactivity: Human, Pig, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR