Anti-Kappa Goat Polyclonal Antibody (AP (Alkaline Phosphatase))

Lieferant: US Biological
I1903-53W
USBII1903-53WEA 665 CHF
USBII1903-53W
Anti-Kappa Goat Polyclonal Antibody (AP (Alkaline Phosphatase))
Antikörper
Recommended dilution: ELISA: 1:50000-1:5000; Western Blot: 1:50000-1:5000; Immunochemistry: 1:500-1:2000
Do not freeze!

Type: Primary
Antigen: Kappa
Clonality: polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR