Anti-IgG, Fc Rabbit Polyclonal Antibody (Rhodamine)

Lieferant: US Biological
I1903-19R7
USBII1903-19R7EA 617 CHF
USBII1903-19R7
Anti-IgG, Fc Rabbit Polyclonal Antibody (Rhodamine)
Antikörper
Type: Primary
Antigen: IgG, Fc
Clonality: polyclonal
Clone:
Conjugation: Rhodamine
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Goat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR