Anti-GFP Rabbit polyclonal antibody

Lieferant: US Biological
G8965-12
USBIG8965-12EA 617 CHF
USBIG8965-12
Anti-GFP Rabbit polyclonal antibody
Antikörper
Recommended dilution: Western Blot: 1:5000; Immunoprecipitation: 1:200

Type: Tag
Antigen: GFP (green fluorescent protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR