Anti-Fluorescein Isothiocyanate Sheep polyclonal antibody

Lieferant: US Biological
F5250-11D
USBIF5250-11DEA 635 CHF
USBIF5250-11D
Anti-Fluorescein Isothiocyanate Sheep polyclonal antibody
Antikörper
Recommended dilution: ELISA: 1:10000-1:40000

Type: Tag
Antigen: Fluorescein Isothiocyanate
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR