Anti-CLASP2 Rabbit polyclonal antibody

Lieferant: US Biological
C5831-03A
USBIC5831-03AEA 937 CHF
USBIC5831-03A
Anti-CLASP2 Rabbit polyclonal antibody
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-1:500

Type: Primary
Antigen: CLASP2 (cytoplasmic linker associated protein 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR