Anti-Calcitonin Gene-Related Peptide Rabbit Polyclonal Antibody

Lieferant: US Biological
C0115-02Y
USBIC0115-02YEA 665 CHF
USBIC0115-02Y
Anti-Calcitonin Gene-Related Peptide Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Calcitonin Gene-Related Peptide
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

TECHNISCHE DATEN

Antigen-Symbol CGRP
Antigen-Name Calcitonin Gene Related Peptide
Environmentally Preferable
Antikörper-Typ Primär
Klonalität Polyklonal
Konjugation Unconjugated
Reaktivität Human
Wirt Rabbit
Isotyp IgG
ELISA Yes
ImmunoChemie Yes
Format Serum
Form lyophilized powder
Lagerungspuffer 50% glycerol, PBS, 10mg/ml BSA.
Lagertemperatur Cool
Transporttemperatur Ice
Validierte Anwendungen Radio Immunoassay

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR