Anti-Calcitonin Gene Related Peptide Rabbit Polyclonal Antibody

Lieferant: US Biological
C0115-02L
USBIC0115-02LEA 1120 CHF
USBIC0115-02L
Anti-Calcitonin Gene Related Peptide Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Calcitonin Gene Related Peptide
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR