Anti-Calcitonin Gene-Related Peptide Chicken Polyclonal Antibody

Lieferant: US Biological
C0115-01G
USBIC0115-01GEA 665 CHF
USBIC0115-01G
Anti-Calcitonin Gene-Related Peptide Chicken Polyclonal Antibody
Antikörper
Recommended dilution: Immunochemistry: 1:100-1:500

Type: Primary
Antigen: Calcitonin Gene-Related Peptide
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR