Anti-BR Serine/Threonine Protein Kinase 1 Rabbit polyclonal antibody unconjugated

Lieferant: US Biological
B2673-75A
USBIB2673-75AEA 888 CHF
USBIB2673-75A
Anti-BR Serine/Threonine Protein Kinase 1 Rabbit polyclonal antibody unconjugated
Antikörper
Recommended dilution: Immunochemistry: 5 ?g/ml

Type: Primary
Antigen: BR Serine/Threonine Protein Kinase 1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR