Anti-ADIPOR1 Rabbit Polyclonal Antibody (Carboxyfluorescein)

Lieferant: US Biological
A0886-75R
USBIA0886-75REA 1630 CHF
USBIA0886-75R
Anti-ADIPOR1 Rabbit Polyclonal Antibody (Carboxyfluorescein)
Antikörper
Type: Primary
Antigen: ADIPOR1 (adiponectin receptor 1)
Clonality: polyclonal
Clone:
Conjugation: Carboxyfluorescein
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR