Anti-APOA4 Goat Polyclonal Antibody

Lieferant: US Biological
151299
USBI151299EA 1140 CHF
USBI151299
Anti-APOA4 Goat Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:32000

Type: Primary
Antigen: APOA4 (apolipoprotein A-IV)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse
Order Now

TECHNISCHE DATEN

Antigen-Symbol APOA4
Environmentally Preferable
Antigen-Name Apolipoprotein A4
Antikörper-Typ Primär
Klonalität Polyklonal
Konjugation Unconjugated
Reaktivität Mouse
Wirt Goat
Isotyp
ELISA Yes
Form liquid
Reinigung purified by immunoaffinity chromatography
Konzentration ~0,5 mg/ml
Lagerungspuffer Tris saline, pH 7,3, 0,5% BSA, 0,02% sodium azide.
Lagertemperatur Cool
Transporttemperatur Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR