Anti-SP-A Rabbit Polyclonal Antibody

Lieferant: US Biological
145210
USBI145210EA 825 CHF
USBI145210
Anti-SP-A Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SP-A
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR