Anti-CDC37 Rabbit Polyclonal Antibody

Lieferant: US Biological
144340
USBI144340EA 825 CHF
USBI144340
Anti-CDC37 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CDC37 (cell division cycle 37)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR