Anti-Listeria sp. Mouse Monoclonal Antibody [clone: 13F145]

Lieferant: US Biological
136687
USBI136687EA 1010 CHF
USBI136687
Anti-Listeria sp. Mouse Monoclonal Antibody [clone: 13F145]
Antikörper
Type: Primary
Antigen: Listeria sp.
Clonality: monoclonal
Clone: 13F145
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR