Anti-UBE2C Mouse Monoclonal Antibody [clone: 9D3]

Lieferant: US Biological
134989
USBI134989EA 920 CHF
USBI134989
Anti-UBE2C Mouse Monoclonal Antibody [clone: 9D3]
Antikörper
Recommended dilution: Immunofluorescence: 10 ?g/ml; Immunochemistry: 3?g/ml

Type: Primary
Antigen: UBE2C (ubiquitin-conjugating enzyme E2C)
Clonality: monoclonal
Clone: 9D3
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR