Anti-ADAT3 Mouse Polyclonal Antibody

Lieferant: US Biological
129154
USBI129154EA 920 CHF
USBI129154
Anti-ADAT3 Mouse Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ADAT3 (adenosine deaminase, tRNA-specific 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR