Anti-CMTM1 Mouse Polyclonal Antibody

Lieferant: US Biological
125113
USBI125113EA 920 CHF
USBI125113
Anti-CMTM1 Mouse Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CMTM1 (CKLF-like MARVEL transmembrane domain containing 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR