Anti-CDK7 Rabbit Polyclonal Antibody

Lieferant: US Biological
124796
USBI124796EA 978 CHF
USBI124796
Anti-CDK7 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CDK7 (cyclin-dependent kinase 7)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR