Anti-AKIP1 Rabbit Polyclonal Antibody

Lieferant: US Biological
123148
USBI123148EA 978 CHF
USBI123148
Anti-AKIP1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: AKIP1 (A kinase (PRKA) interacting protein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR