Anti-NONO Rabbit Polyclonal Antibody (Biotin)

Lieferant: US Biological
039054-BIOTIN
USBI039054-BIOTINEA 1250 CHF
USBI039054-BIOTIN
Anti-NONO Rabbit Polyclonal Antibody (Biotin)
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:10-50

Type: Primary
Antigen: NONO (non-POU domain containing, octamer-binding)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR