Anti-NDN Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Lieferant: US Biological
038844-FITC
USBI038844-FITCEA 1250 CHF
USBI038844-FITC
Anti-NDN Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikörper
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500; Flow Cytometry: 1:10-50
Do not freeze!

Type: Primary
Antigen: NDN (necdin, melanoma antigen (MAGE) family member)
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR