Anti-MARCH5 Rabbit Polyclonal Antibody (APC (Allophycocyanin))

Lieferant: US Biological
038196-APC
USBI038196-APCEA 1250 CHF
USBI038196-APC
Anti-MARCH5 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Antikörper
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:50-100; Flow Cytometry: 1:10-50
Do not freeze!

Type: Primary
Antigen: MARCH5 (membrane-associated ring finger (C3HC4) 5)
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR