Anti-KCND3 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))

Lieferant: US Biological
037307-AP
USBI037307-APEA 1250 CHF
USBI037307-AP
Anti-KCND3 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500
Do not freeze!

Type: Primary
Antigen: KCND3 (potassium voltage-gated channel, Shal-related subfamily, member 3)
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR