Anti-KAT2A Rabbit Polyclonal Antibody (APC (Allophycocyanin))

Lieferant: US Biological
037273-APC
USBI037273-APCEA 1250 CHF
USBI037273-APC
Anti-KAT2A Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Antikörper
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!

Type: Primary
Antigen: KAT2A (K(lysine) acetyltransferase 2A)
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR