Anti-GPR149 Rabbit Polyclonal Antibody (PE (Phycoerythrin))

Lieferant: US Biological
036225-PE
USBI036225-PEEA 1250 CHF
USBI036225-PE
Anti-GPR149 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Antikörper
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!

Type: Primary
Antigen: GPR149 (G protein-coupled receptor 149)
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR