Anti-ATAD3C Rabbit Polyclonal Antibody (Biotin)

Lieferant: US Biological
032155-BIOTIN
USBI032155-BIOTINEA 1250 CHF
USBI032155-BIOTIN
Anti-ATAD3C Rabbit Polyclonal Antibody (Biotin)
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: ATAD3C (ATPase family, AAA domain containing 3C)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR