Anti-Kappa light chain Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: HP6053]

Lieferant: US Biological
031066-FITC
USBI031066-FITCEA 1250 CHF
USBI031066-FITC
Anti-Kappa light chain Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: HP6053]
Antikörper
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-1000; Immunochemistry: 1:10-50

Type: Primary
Antigen: Kappa light chain
Clonality: Monoclonal
Clone: HP6053
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR