Laser Protection Goggles
Lieferant: UNIVET
562S.00.01.312
UNVT562S.00.01.312EA 563 CHF
UNVT562S.00.01.312
Laser Protection Goggles
Schutzbrillen
  • Filter: Full protection
  • VLT: 40%
  • Alignement laser wavelength T%(λ) >10%: 410 to 695 nm