Supelco® Organic Reference Standard, Fatty Acids Unsaturated Kit

Lieferant: Merck

UN10-1KT
UN10-1KTEA 585 CHF
UN10-1KT
Supelco® Organic Reference Standard, Fatty Acids Unsaturated Kit
Standards Organische Standards

  • GHS signal: GHS07


Lieferumfang: Analytes: Arachidonic acid 100 mg; cis-4,7,10,13,16,19-Docosahexaenoic acid 100 mg; Elaidic acid 100 mg; Erucic acid 100 mg; Linoleic acid 100 mg; Linolenic acid 100 mg; Nervonic acid 100 mg; Oleic acid 100 mg; Palmitoleic acid 100 mg; Petroselinic acid 100 mg.


Bitte beachten P261 - P264 - P271 - P273 - P302 + P352 - P305 + P351 + P338

Order Now

TECHNISCHE DATEN

M-Clarity Qualitätsstufe MQ100
Environmentally Preferable
Beschreibung Gefahrenklasse H315 - H319 - H335 - H413
Flammpunkt 62 °C
Shipping conditions DICE
Lagerbedingungen –20 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR