Cell culture dishes with gripping ring
Lieferant: TPP
93100 93150 93040 93060 393040 393100
TPPA93100EA 222 CHF
TPPA93100 TPPA93150 TPPA93040 TPPA93060 TPPA393040 TPPA393100
Cell culture dishes with gripping ring
Zellkulturschalen Zell- und Gewebekultur-Platten
These PS cell culture dishes are designed for surface activation of the growth area enhancing optimal proliferation of cells.
  • Designed for manual handling
  • Side walls of dish not surface activated
  • Serrated edges of gripping ring prevent accidental dropping of the dish bottom
  • Lateral yellow inscription field on the lid
  • Spacer on the inside of the dish lid provide constant air flow
  • Numeric 'clock' markings (12, 3, 6, 9) in the base of the dish divide it into quadrants for quick orientation
Vents in the dish base allow air to flow between stacked dishesMore Product Information