Industrial Vacuum Cleaner, C-35L EX (DT) MFS
Lieferant: TIGER-VAC
111783B
TIGV111783BEA 0 CHF
TIGV111783B
Industrial Vacuum Cleaner, C-35L EX (DT) MFS
Staubsauger
Industrial vacuum cleaner for continuous duty. Designed specifically for housekeeping and general cleaning. It is ideal for general cleaning and for process integration.
  • Static dissipating
  • HEPA/SD accessories - included
  • High filtration efficiency unit
  • Stainless general
  • Dry recovery and detachable tank
  • Brushless motor available
  • By-pass Ametek® long life motor
Longer life brushless motor available. Always with 4×H14 HEPA filters, Manual Filter Shaker (MFS) with static dissipative teflon laminated main cloth filter assembly, Detachable recovery tank (DT)More Product Information