Cleanroom wipes, FoamWipe®, TX704
Lieferant: Texwipe
TX704
TEXWTX704EA 1280 CHF
TEXWTX704
Cleanroom wipes, FoamWipe®, TX704
Wischtücher Reinraum-Wischtücher Trockene Reinraumwischtücher
These FoamWipe™ wipes are made from open-cell, 100 pore-per-inch polyurethane foam.
  • Silicone free
  • Precision cut for low particle generation
  • Can be used as soft and nonabrasive work surface
They are an excellent replacement for cellulose sponges in critical environments.