Wall mounted scrub sinks
Lieferant: TEKNOMEK
WSSS-1SIB WSSS-1SIM WSSS-1SKO WSSS-1SLT WSSS-2SIB WSSS-2SIM WSSS-2SKO WSSS-2SLT
TEKMWSSS-1SIBEA 0 CHF
TEKMWSSS-1SIB TEKMWSSS-1SIM TEKMWSSS-1SKO TEKMWSSS-1SLT TEKMWSSS-2SIB TEKMWSSS-2SIM TEKMWSSS-2SKO TEKMWSSS-2SLT
Wall mounted scrub sinks
Armaturen und Spülbecken Spülbecken
Our wall mounted scrub sink is a perfect solution for veterinary practices with a variety of different operations- lever, knee paddle or infra-red sensor
  • Brushed dull polish finish
  • Various spout operations available
  • Battery or mains powered infrared taps with a spout length of 250 mm
  • Lever tap option comes with 150 mm levers
  • Deep bowl to enable scrubbing to elbow
  • Hygienic design to prevent water pooling
Our scrub sink is manufactured from 304 food-grade stainless steel and is available in 1 and 2 stations with a wide range of tap operations; kneeMore Product Information