2'-Desoxyadenosin Monohydrat ≥99,0% (durch titrimetrische Analyse)

Lieferant: TCI

Synonyms: (2R,3S,5R)-5-(6-Amino-9H-purin-9-yl)-2-(hydroxymethyl)tetrahydrofuran-3-ol hydrate

D0046-25G D0046-5G
TCIAD0046-25GEA 888 CHF
TCIAD0046-25G TCIAD0046-5G
2'-Desoxyadenosin Monohydrat ≥99,0% (durch titrimetrische Analyse)
2'-Desoxyadenosin Monohydrat
Formel: C₁₀H₁₃N₅O₃·1H₂O
MW: 269.26 g/mol (251.24 anhy)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00149364
CAS-Nummer: 16373-93-6
EINECS: 213-488-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8db019abbbcffd458a3267a4cae6fba266261d87&vid=8db019abbbcffd458a3267a4cae6fba266261d87&prid=web&clid=SAPDAM

Order Now

Spezifikation Testergebnisse

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 99.0 % (calcd.on anh.substance)
Specific rotation [a]20/D -24.0 to -27.0 deg (C=0.5, H2O)
Solubility in hot Water almost transparency
Water 5.0 to 7.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR