Spark® Cyto multimode plate readers with live cell imaging and real-time cytometry
Lieferant: Tecan
94000004 94100004 94200004 94300004 94400004
TCAT94000004EA 0 CHF
TCAT94000004 TCAT94100004 TCAT94200004 TCAT94300004 TCAT94400004
Spark® Cyto multimode plate readers with live cell imaging and real-time cytometry
Mikrotiterplatten-Photometer
Spark Cyto is the first multimode reader with live cell imaging and real time cytometry to offer as much information as possible from the same cells, under the same conditions.
  • Live cell imaging and real-time cytometry for 2D and 3D cell models
  • Temperature and gas control
  • Lid Lifter™
  • Enhanced Absorbance, Fluorescence and Luminescence modules
  • Fusion optics - combine the flexibility of monochromators with the sensitivity of filters
  • ALPHA technology
  • Fluorescence filter top/bottom
  • DNA-, RNA- & protein quantification
  • Low sample volume: 2 µl
  • Up to 16 sample
More Product Information