Tetramethylrhodamine methyl ester perchlorate (TMRM) ≥95% (durch HPLC) geeignet für Fluoreszenz, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY

Synonyms: N-[6-(Dimethylamino)-9-[2-(methoxycarbonyl)phenyl]-3H-xanthen-3-ylidene]-N-methylmethanaminium Perchlorate, Tetramethylrhodamine methyll ester Perchlorate, 3,6-Bis(dimethylamino)-9-(2-(methoxycarbonyl)phenyl)xanthylium perchlorate, Tetramethylrhodamine Methyl Ester Perchlorate

M-Clarity™ quality level = MQ200

T5428-25MG
T5428-25MGEA 460 CHF
T5428-25MG
Tetramethylrhodamine methyl ester perchlorate (TMRM) ≥95% (durch HPLC) geeignet für Fluoreszenz, Sigma-Aldrich®
Tetramethylrhodamine methyl ester perchlorate (TMRM)

Tetramethylrhodamine methyl ester perchlorate (TMRM) is a fluorescent dye that can label mitochondria and mark embryonic stem cell/induced pluripotent stem cell-derived cardiomyocytes. It is useful for the isolation of mature cardiomyocytes.


  • Fluorescence - λem 573 nm in methanol


Tetramethylrhodamine methyl ester perchlorate (TMRM) has been used as a sensitive probe for the detection of mitochondrial membrane potential.

Formel: C₂₅H₂₅N₂O₃·ClO₄
Lager Temperatur: Kühlschrank
MDL Number: MFCD00274460
CAS-Nummer: 115532-50-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d060469fe51a4f584a9ed99743f66e0241db594f&vid=a9086ff23eac973df6d3dc7e73aa2f47746173f2&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR