ACM craft tools
Lieferant: SWANN MORTON
9101 9105 9148 9144 9142 9138 9137 9136 9131 9130 9127 9122 9107 9106 9303 9307 9308 9311
SWAN9101EA 230 CHF
SWAN9101 SWAN9105 SWAN9148 SWAN9144 SWAN9142 SWAN9138 SWAN9137 SWAN9136 SWAN9131 SWAN9130 SWAN9127 SWAN9122 SWAN9107 SWAN9106 SWAN9303 SWAN9307 SWAN9308 SWAN9311
ACM craft tools
Skalpelle Skalpelle, komplett
The ACM tool set is presented in a resilient blue carry case and contains a comprehensive range of 13 precision ground carbon steel blades manufactured to a high surgical quality.
Also including three handle variations the set can be used for general cutting applications including the trimming of light and medium weight materials, precision trimming, stencilling, scoring and etching as well as corner cuts, deburring, stripping and gasket cutting making this set extremely versatile.
ACM No. 1 handle - for light duty applications
A light duty craft knife designed to assist with smallerMore Product Information